Learn More
Cayman Chemical Bafilomycin A1, 88899-55-2, 500 µg
SDP
Supplier:  Cayman Chemical 11038500
Encompass_Preferred
A selective, reversible inhibitor of vacuolar H+-ATPases (V-ATPases), blocking these proton pumps in mammalian, plant, or fungal cells with an IC50 value in the 4-400 nM range; also inhibits autophagy by preventing vacuolar acidification necessary for autophagosome maturation
Catalog No. NC0910155
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.