Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Bafilomycin A1, (V)-ATPase Inhibitor, Solid, 10 μg

Catalog No. 502584589
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4589 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4589 Supplier Abcam Supplier No. AB12049710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly potent, selective vacuolar (V)-ATPase inhibitor that induces vacuolar deacidification and promotes apoptosis.

  • Molecular Weight: 622.8 Da
  • Inhibitor: Highly potent, selective vacuolar (V)-ATPase inhibitor (IC50 = 500 pM in chromaffin granule membranes)
  • Biological Effect: Induces vacuolar deacidification and promotes apoptosis
  • Form: Solid
  • CAS Number: 88899-55-2
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule V-ATPase
Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Formulation 100% Bafilomycin A1
Quantity 10 μg
For Use With (Application) Vacuolar (V)-ATPase inhibition, Vacuolar deacidification, Apoptosis induction
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.