Learn More
AdipoGen Bafilomycin B1,Chemical. CAS: 88899-56-3
SDP
Supplier:  AdipoGen BVT0004M001
Encompass_Preferred
Bafilomycin B1, AdipoGen Life Sciences. Chemical. CAS: 88899-56-3. Formula: C44H65NO13. MW: 815.9. Isolated from Streptomyces hygroscopicus. Macrolide antibiotic. Specific vacuolar-type H+-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Antitrypanosomal and antileishmanial compound. Phytotoxic. Influences the cell wall biosynthesis of Asp. niger. 88899-56-3
Catalog No. 50-117-6187
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.