Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAG 956

Catalog No. 360610R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-061-0R 10 mg
36-065-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-061-0R Supplier Tocris Bioscience™ Supplier No. 3606/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Dual PI 3-kinase and PDPK1 (PDK1) inhibitor

BAG 956 is a dual PDPK1 (PDK1) and class I PI 3-K inhibitor (IC50 values are 245, 56, 446, 35 and 117 nM for PDPK1 and PI 3-K p110 -α, -β, -δ, and -γ respectively). Inhibits cellular AKT phosphorylation at Thr308. Blocks cell proliferation, causing arrest in G1 phase of the cell cycle. Slows tumor progression in mouse xenograft models.

Specifications

Purity 98%
Content And Storage Store at -20°C
Quantity 10 mg
Product Type BAG 956
Molecular Weight (g/mol) 427.5
Inhibitors BAG 956

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.