Learn More
Andwin Scientific BAG CLAVIES PP 24X36IN PK100
SDP
Supplier:  Andwin Scientific 95042558
Encompass_Preferred
With the included twist ties, the autoclavable polypropylene bags feature secure closure and proper ventilation during the process of sterilization. This product is also ideal for autoclaving hazardous and nonhazardous materials and waste. The bags comply with the strength requirements in ASTM D 1709-04 for 165g dart drop test and work with autoclaving temperatures up to 135C (275F).
Catalog No. NC0092682
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.