Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAGE4 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501732682 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2682 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2682 Supplier Proteintech Group Inc Supplier No. 200711AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody can recognize BAGE4, BAGE, BAGE3, BAGE5.

BAGE4, also named as MLL3P and CT2.4, belongs to the BAGE family. It is a candidate gene encoding tumor antigens. BAGE4 is Cancer/testis antigen 2.4.

Specifications

Antigen BAGE4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BAGE3
Gene Accession No. Q13072, Q86Y27, Q86Y28, Q86Y29
Gene Alias B melanoma antigen 4, BAGE4, Cancer/testis antigen 2.4, CT2.4, MLL3P
Gene Symbols BAGE, BAGE3, BAGE4, BAGE5
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 574, 85316, 85317, 85318
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.