Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baker-flex™, Cellulose F, J.T. Baker™

Catalog No. 02002560 Shop All Avantor Products
Change view
Click to view available options
Quantity:
50 Sheets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-560 50 Sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-002-560 Supplier Avantor Supplier No. 446900
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible TLC sheet coated with high-purity cellulose powder and a fluorescent indicator for chromatography applications.

Flexible TLC Sheet

Chemical Identifiers

CAS 9004-34-6

Specifications

Color White
Physical Form Crystal or Powder
Quantity 50 Sheets
Odor Odorless
Solubility Information Insoluble in water
Chemical Name or Material Cellulose F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.