Learn More
Description
Flexible TLC Sheet
Specifications
Specifications
| Color | White |
| Physical Form | Crystal or Powder |
| Quantity | 25 Sheets |
| Odor | Odorless |
| Solubility Information | Insoluble in water |
| Chemical Name or Material | Cellulose Microcrystalline |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.