Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avantor™ BAKERBOND spe™ Silica Gel (SiOH) Disposable Extraction Columns, J.T. Baker™
Supplier: Avantor™ 708626
This item is not returnable.
View return policy
Specifications
6 mL | |
Disposable extraction column | |
Granules | |
Silica | |
Pack of 250 |
Silica gel | |
500 mg | |
40 μm | |
60 angstroms | |
White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction