Learn More
Sartorius Balance Interface Cables
Catalog No. 01913353
Change view
Click to view available options
Cable Interface D1 To Ibm At
Cable Interface D1 To Ibm Pc
Cable25pin-rj11 6ft For Xl Bal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-913-353 Cable25pin-rj11 6ft For Xl Bal
01-913-370 Cable Interface D1 To Ibm Pc
01-913-371 Cable Interface D1 To Ibm At
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-913-353 Supplier Sartorius Supplier No. 600324.1
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Interface Cables. Connect your Sartorius balance to a 9-pin connector on a computer. All cables are 1m long.