Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Balance and Scale Weighing Pans

Catalog No. 14557551 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Cubis I Lab Balance
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-557-551 Cubis I Lab Balance
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-557-551 Supplier Sartorius Supplier No. YWP01MS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To prevent any damage to the surface of weights, Sartorius offers you a wide range of accessories for the proper handling, storage and cleaning of weights.

Weighing pans for use with Sartorius Balances.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Bottom Shape Flat
Color Silver
Disposable Reusable
Material Stainless Steel
Shape Round
Sterility Non-sterile
Type Weighing Pan
For Use With (Application) Weighing samples
For Use With (Equipment) Cubis I Lab Balance
Diameter (English) 5.1 in.
Diameter (Metric) 130 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.