Learn More
Anton Paar Set of 5 stainless steel balls 25 mm
Supplier:  Anton Paar 177691
Encompass_Preferred
Grinding ball with 25 mm diameter made of INOX for use with grinding jars made of INOX.Suitable for use with Anton Paar BM500 Ball Mill NC1818284 and Grinding Jar NC1818286 or NC1818288
Catalog No. NC1818289