Description
- Metallic-coated ball on the end of a thread is attracted by induction until it touches a Van de Graaff generator dome
- Violently repels, flying around the dome without touching it
- Silver snake does not retain its charges and repeatedly strikes back at the generator dome
Specifications
Specifications
| For Use With (Application) | Great for the classroom |
| For Use With (Equipment) | Van de Graaff generator |
| Product Type | Ball and Snake Demo |
| Material | Metallic |