Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bal Supply Ball and Socket Joint Clips, Keck-Type

Catalog No. 13684170 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Diameter (Metric) Inside:
13 mm
19 mm
29 mm
35 mm
Color:
Blue
Orange
Red
Violet
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inside Color
13-684-170 29 mm Red
13-684-168 13 mm Violet
13-684-169 19 mm Blue
13-684-171 35 mm Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 13-684-170 Supplier Bal Supply Supplier No. 203229
Add to Cart
Add to Cart
This item is not returnable. View return policy

Made from polyoxymethylene (POM), Bal Supply Keck-Type Ball and Socket Joint Clips are color-coded for easy size identification.

  • POM construction will not scratch glass
  • For use with ball and socket joints

Specifications

Autoclavable Autoclavable
Quantity 10 Pk.
Type Keck Joint Clip
Material POM
For Use With (Application) For Ball and Socket Joints
Color Red
Diameter (Metric) Inside 29 mm
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.