Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Ballistics Car
SDPSCI_ED

Catalog No. S139498 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139498 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139498 Supplier United Scientific™ Supplier No. BLCAR1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Classic visual demonstration that will capture student's attention and interest.

  • Push the cart to eject the steel ball from the spring-loaded barrel
  • The ball falls back into the barrel of the moving car, demonstrating that the forward motion of the ball is the same as the vehicle from which it was ejected
  • Comes with a steel ball, release pin, and cord
  • The barrel permits two different heights of ball flight
  • Includes activity guide

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.