Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Balloons for Science Kits
SCI_ED

Catalog No. S05151 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (English) Outer:
7 in.
Assorted
Shape:
Oblong
Round
Color:
Assorted
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Outer Shape Color
S05151 7 in. Round -
S05150 Assorted Oblong Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S05151
This item is not returnable. View return policy

Consumbles for use with multiple projects and science kits.

  • Round
  • Oblong

Specifications

Product Type Balloon
For Use With (Application) For use in multiple projects and in science kits
Shape Round
Diameter (English) Outer 7 in.
Diameter (Metric) Outer 17.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.