Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Balofloxacin Dihydrate 98.0+%, TCI America™
SDP

Catalog No. B43541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43541G 1 g
B43545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43541G Supplier TCI America Supplier No. B43541G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 151060-21-8
Molecular Formula C20H28FN3O6
Molecular Weight (g/mol) 425.457
MDL Number MFCD00864925
InChI Key WEGNYJXOCPJAPS-UHFFFAOYSA-N
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[3-(methylamino)-1-piperidyl]-4-oxoquinoline-3-carboxylic Acid
PubChem CID 6918202
IUPAC Name 1-cyclopropyl-6-fluoro-8-methoxy-7-[3-(methylamino)piperidin-1-yl]-4-oxoquinoline-3-carboxylic acid;dihydrate
SMILES CNC1CCCN(C1)C2=C(C=C3C(=C2OC)N(C=C(C3=O)C(=O)O)C4CC4)F.O.O

Specifications

Melting Point 135°C
Color White
Quantity 1 g
Formula Weight 389.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Balofloxacin Dihydrate
RTECSNumber VB1993755
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.