Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bamlanivimab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542311
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIMA542311 100 μg
1 options
This item is not returnable. View return policy

Catalog No. PIMA542311

Supplier: Invitrogen™ MA542311

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Bamlanivimab is a biosimilar that targets RBD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bamlanivimab Humanized
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias LY3819253; LY-CoV555
Host Species Human
Immunogen SARS-CoV-2 RBD.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.