Learn More
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human SLC4A1 shares 75.7% and 74.5% amino acid (aa) sequence identity with and rat SLC4A1, respectively. Positive Control - WB: human K562 whole cell. IHC: human spleen tissue. Flow: HepG2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | Band 3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 5G2G7 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and no preservative |
| Gene | SLC4A1 |
| Gene Accession No. | P02730 |
| Gene Alias | AE 1; AE1; anion exchange protein 1; anion exchanger 1; anion exchanger-1; band 3 anion transport protein; BND3; CAMP; CD233; CHC; DI; Diego blood group; EMPB3; EPB3; erythrocyte membrane protein band 3; erythroid anion exchange protein; FR; Froese blood group; l11Jus51; MEB3; MGC116750; MGC116753; MGC126619; MGC126623; RTA1A; SAO; Slc4a1; solute carrier family 4 (anion exchanger), member 1; solute carrier family 4 (anion exchanger), member 1 (Diego blood group); solute carrier family 4 member 1; solute carrier family 4 member 1 (Diego blood group); solute carrier family 4, anion exchanger, member 1 (erythrocyte membrane protein band 3, Diego blood group); solute carrier family 4, anion exchanger, number 1; Solute carrier family 4, member 1, anion exchange protein 1 (kidney band 3); SPH4; SW; Swann blood group; Waldner blood group; WD; WD1; WR; Wright blood group; ZNF828 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
