Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bandmate™ 4L Nalgene™ Bottle

Catalog No. BW0010
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW0010 1 bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BW0010 Supplier Invitrogen™ Supplier No. BW0010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Bandmate 4L Nalgene Bottle is a replacement bottle for use with the Bandmate Automated Western Blot Processor.

The bottle is used for storing wash buffer used throughout western blot processing. Each package contains one 4L bottle with quick connection tubing.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description 4L Nalge Bottle
For Use With (Application) Western Blot
For Use With (Equipment) Invitrogen Bandmate Automated Western Blot Processor
Product Line Bandmate
Type Bottle
Quantity 1 bottle
Shipping Condition Ambient
Dimensions (L x W x H) 25.654 x 21.59 x 6.045 cm
Weight 0.227 kg
Weight (English) 8 oz
Weight (Metric) 0.227 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.