Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bandmate™ Reagent Tubes

Catalog No. BW020X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 tubes
500 tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BW020X5 100 tubes
BW020X25 500 tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BW020X5 Supplier Invitrogen™ Supplier No. BW020X5
Add to Cart
Add to Cart

Bandmate Reagent Tubes are specially designed for use with the Bandmate Automated Western Blot Processor.

The tubes are required for dispensing primary and secondary antibodies into the Bandmate mini and midi incubation trays during the automated western blot processing procedure. Two reagent tubes are required for each blot.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description Bandmate Reagent Tubes, 100 Pack
For Use With (Application) Western Blot
For Use With (Equipment) Invitrogen Bandmate Automated Western Blot Processor
Product Line Bandmate
Type Reagent Tube
Quantity 100 tubes
Shipping Condition Ambient
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.