Learn More
Invitrogen™ Bandmate™ Reagent Tubes
Description
Specifications
Specifications
| Color | Clear |
| Description | Bandmate Reagent Tubes, 500 Pack |
| For Use With (Application) | Western Blot |
| For Use With (Equipment) | Invitrogen Bandmate Automated Western Blot Processor |
| Product Line | Bandmate |
| Type | Reagent Tube |
| Quantity | 500 tubes |
| Shipping Condition | Ambient |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.