Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BANK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557910 Supplier Invitrogen™ Supplier No. PA557910
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RHGHKELKKI FEDFSIQEID INNEQENDYE EDIASFSTYI PSTQNPAFHH ESRKTYGQSA DGAEANEMEG EGKQNGSGME TKHSPL Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 55%.

The protein encoded by this gene is a B-cell-specific scaffold protein that functions in B-cell receptor-induced calcium mobilization from intracellular stores. This protein can also promote Lyn-mediated tyrosine phosphorylation of inositol 1,4,5-trisphosphate receptors. Polymorphisms in this gene are associated with susceptibility to systemic lupus erythematosus. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BANK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BANK1
Gene Accession No. Q8NDB2
Gene Alias A530094C12Rik; AI451642; AVIEF; B cell scaffold protein with ankyrin repeats 1; BANK; Bank1; B-cell scaffold protein with ankyrin repeats; B-cell scaffold protein with ankyrin repeats 1; B-cell scaffold protein with ankyrin repeats 1 delta 2 transcript; LRRGT00104; Protein AVIEF
Gene Symbols BANK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BANK1. Recombinant protein control fragment (Product #RP-96695).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55024
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.