Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520499
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520499 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520499 Supplier Invitrogen™ Supplier No. PA520499
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-20499 can be used with blocking peptide PEP-0619.

BAP3 was initially identified through interaction in a yeast two-hybrid system with the brain-specific angiogenesis inhibitor 1, a p53-target gene that encodes a seven-span transmembrane protein member of the secretin receptor family. BAP3 is predominantly expressed in the brain and possess high homology with Munc13 and synaptotagmin, suggesting that BAP3 may play a role in regulating neurotransmitter release. Recent experiments have shown that BAP3 is induced in certain tumors such as desmoplastic small round cell tumor. Ectopic expression of BAP3 in tumor cells dramatically enhances growth in low serum conditions and colony formation in soft agar, suggesting that the regulated exocytotic pathway may play a role in cancer cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAP3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene BAIAP3
Gene Accession No. O94812
Gene Alias AL031709.1; BAI1 associated protein 3; BAI1-associated protein 3; BAIAP3; BAI-associated protein 3; BAP3; Brain-specific angiogenesis inhibitor I-associated protein 3; Gm937; KIAA0734
Gene Symbols BAIAP3
Host Species Rabbit
Immunogen A 13 amino acid peptide from near the amino terminus of human BAP3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8938
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.