Learn More
Apexbio Technology LLC Oxytocin acetate 6233-83-6 10mg
SDP
Supplier:  Apexbio Technology LLC B472310
Encompass_Preferred
Oxytocin acetate (CAS 6233-83-6) is an acetate salt form of oxytocin a neuropeptide synthesized primarily in hypothalamic regions including the paraventricular and supraoptic nuclei Biologically oxytocin acts as a positive allosteric modulator of the -opioid receptor (MOR) enhancing receptor-mediated signaling without directly activating MOR or altering opioid ligand affinity In vitro studies with hMOR-expressing HEK293 cells indicate oxytocin augments signaling induced by MOR agonists such as endomorphin-1 morphine and fentanyl Its intracerebroventricular administration increases neuronal activity in rat nucleus accumbens shell highlighting potential roles in pain modulation social behavior and addiction research
Catalog No. 50-000-62846
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.