Learn More
Med Vet International Corid, (Amprolium) 9.6%, Oral Solution, 16oz
Supplier:  Med Vet International PHVCORIDOS
Encompass_Preferred
Coccidiostat (amprolium) that aids in the treatment and prevention of coccidiosis caused by Eimeria bovis and E. zurnii in calves.
Catalog No. 50-282-8817