Learn More
Cole-Parmer Hose Barb Fitting, Straight Reducer, Stainless Steel, 5/32" ID x 3/32" ID; 10/Pk
Supplier:  Cole-Parmer 5010871
Encompass_Preferred
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. Use the straight reducers to connect two different size tubing. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
Catalog No. 50-255-8356