Learn More
StatLab Bard-Parker Blade Handle
SDP
Bard-Parker Blade Handle
$71.70 - $96.10
Specifications
Disposable Reusable
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
22-000-016
SDP
View Documents
StatLab
BP1060
No. 6
Each for $71.70
Only null left
Add to Cart
22-000-017
SDP
View Documents
StatLab
BP1080
No. 8
Each for $96.10
Only null left
Add to Cart
Description
Description
  • Intended to be used with Bard-Parker conventional blades - BP1322 and BP1340
  • High-impact poly handles are designed for a contoured, firm grip and easy manipulation
Specifications
Specifications
Reusable
Non-sterile
Not Autoclavable
No
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.