Learn More
StatLab Bard-Parker Scalpel Blade
SDP
Bard-Parker Scalpel Blade
$323.50 - $804.00
Specifications
Product Type Blade
Disposable Single-use
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
22-000-018
SDP
View Documents
StatLab
BP1322
# 22
Case of 150 for $323.50
Only null left
Add to Cart
22-000-019
SDP
View Documents
StatLab
BP1340
# 60
Case of 150 for $804.00
Only null left
Add to Cart
Description
Description
  • Bard-Parker rib-back scalpel blades offer the superior cutting advantages of carbon steel technology
  • The exclusive Rib-Back blade design for greater strength and control
  • Specially ground and carefully inspected
  • Packaged 150 per box, non-sterile and individually wrapped
Specifications
Specifications
Blade
Non-sterile
No
Single-use
Not Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.