Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Description
Offer consistent sharpness, control and strength
- Each blade packaged in foil pouch with liner for maximum protection of blade and user
- Rib back blade design on select blades offers greater cutting control and strength
- Available with plastic shield SafetyLock to securely cover the blade edge before and after use
- Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
Specifications
| Blade Material | Carbon Steel |
| Product Type | Surgical Blade |
| For Use With (Application) | For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut |
| Handle Type | Ribbed |
| Size | # 21 |
| Sterility | Non-sterile |