Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Offer consistent sharpness, control and strength
Supplier: Aspen Surgical™ 371156
This item is not returnable.
View return policy
Description
Offer consistent sharpness, control and strength
- Each blade packaged in foil pouch with liner for maximum protection of blade and user
- Rib back blade design on select blades offers greater cutting control and strength
- Available with plastic shield SafetyLock to securely cover the blade edge before and after use
- Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
Carbon Steel | |
For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut | |
# 22 |
Surgical Blade | |
Safetylock Ribbed | |
Sterile |