Learn More
Aspen Surgical™ Bard-Parker™ Surgical Blades
Offer consistent sharpness, control and strength
Supplier:  Aspen Surgical™ 371315
 View more versions of this product
Catalog No. 08-915-1A
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Offer consistent sharpness, control and strength
  • Each blade packaged in foil pouch with liner for maximum protection of blade and user
  • Rib back blade design on select blades offers greater cutting control and strength
  • Available with plastic shield SafetyLock to securely cover the blade edge before and after use
  • Available sterile in carbon and stainless steel and nonsterile in carbon steel
Specifications
Specifications
Carbon Steel
For Tissue Separation, and Other Procedures that require a Sharp Surgical Blade to Puncture or Cut
# 15
Surgical Blade
Ribbed
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.