Learn More
Selleck Chemical LLC Bardoxolone 5mg 218600-44-3 CDDO, RTA 401
SDP
Supplier:  Selleck Chemical LLC S66475MG
Encompass_Preferred
Bardoxolone (CDDO, RTA 401), acts by releasing Nrf2 from KEAP1,is a highly potent activator of Nrf2 that induce programmed cell death (apoptosis) in cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1819830
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.