Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium 2,4-pentanedionate

Catalog No. AA1461314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1461314 25 g
AA1461322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1461314 Supplier Thermo Scientific Chemicals Supplier No. 01461314
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12084-29-6 | C10H14BaO4 | 335.55 g/mol

Barium 2,4-pentanedionate hydrate is used in the preparation of radioluminescence of poly ethylene glycolated Eu3+ doped nanophosphors as bioimaging probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12084-29-6
Molecular Formula C10H14BaO4
Molecular Weight (g/mol) 335.55
MDL Number MFCD00013484
InChI Key DJHZYHWLGNJISM-FDGPNNRMSA-L
Synonym barium 2+ ; z-4-oxopent-2-en-2-olate, barium 2,4-pentanedionate, bis pentane-2,4-dionato-o,o' barium, barium, bis 2,4-pentanedionato-o,o'-, t-4, 2,4-pentanedione,ion 1-, barium 2:1, barium, bis 2,4-pentanedionato-o,o'-, hydrate
PubChem CID 5486157
SMILES [Ba++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point >320°C
Physical Form Powder
Quantity 25 g
UN Number UN1564
Solubility Information Insoluble in water.
IUPAC Name barium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 335.56 (Anhydrous)
Chemical Name or Material Barium 2,4-pentanedionate hydrate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 235-151-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.