Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, 1.50 M, Ricca Chemical
SDP

Catalog No. R0882800500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0882800500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0882800500 Supplier Ricca Chemical Company Supplier No. R0882800500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compendial reagent for TAPPI analysis of cellulose.

Chemical Identifiers

CAS 7732-18-5 , 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Color Colorless
CAS Min % 72.0, 26.48
CAS Max % 74.0, 27.56
Physical Form Liquid
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1.5 Normal
Odor Odorless
Packaging Natural Poly Bottle
Chemical Name or Material Barium Chloride
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.