Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, 10% w/v, Aqueous, Neutralized, Reagents

Catalog No. 01335823
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-823 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-823 Supplier Reagents Holdings Llc Supplier No. CB1003004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barium Chloride, 10% w/v, Aqueous, Neutralized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10326-27-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Colorless
pH 7
Physical Form Liquid
Quantity 4 L
Percent Purity 10% w/v
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.092
Chemical Name or Material Barium Chloride, 10% w/v, Aqueous, Neutralized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.