Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, Anhydrous, 98%, Spectrum™ Chemical
SDP

Catalog No. 18608981 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-981 125 g Poly Bottle
18-608-983 500 g Poly Bottle
18-608-984 2.5 kg Poly Bottle
18-608-982 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-981 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1028125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10361-37-2

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 125 g
Percent Purity 98%
Grade Ungraded
Packaging Poly Bottle
Chemical Name or Material Barium Chloride, Anhydrous

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.