Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium chloride dihydrate, 99+%, for analysis

Catalog No. p-3734649
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC203135000 500 g Plastic bottle
AC203130010 1 kg Plastic bottle
AC203130025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC203135000 Supplier Thermo Scientific Chemicals Supplier No. 203135000
Only null left
Add to Cart

CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560.0°C
Melting Point 962.0°C
Color Gray-White
pH 5.2 to 8.2 (5% soln. at 25°C)
Quantity 500 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula BaCl2·2H2O
Merck Index 15, 967
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, practically insoluble in ethanol, acetone and, ethyl acetate
Formula Weight 244.28
Percent Purity ≥99%
Grade Analytical
Loss on Drying 14.0 % to 16.0 % (150°C)
Packaging Plastic bottle
Chemical Name or Material Barium chloride dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
If eye irritation per

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.