Learn More
Barium chloride dihydrate, 99+%
Description
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
Notes
Stable and hygroscopic in nature.
Specifications
Specifications
| Melting Point | 113°C (in 2H2O) |
| Quantity | 1000 g |
| Odor | Odorless |
| Density | 3.097 |
| UN Number | UN1564 |
| Merck Index | 14,971 |
| Solubility Information | Soluble in water and metanol. Insoluble in ethanol and ethyl acetate. |
| IUPAC Name | barium(2+);dichloride;dihydrate |
| Formula Weight | 244.28 (208.25 Anhydrous) |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
| Hazard Category | H301-H319-H332 |
| Hazard Statement | GHS H Statement H301-H331 Toxic if swallowed. Toxic if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S. |
| EINECSNumber | 233-788-1 |
| RTECSNumber | CQ8751000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.