Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium chloride dihydrate, ACS

Catalog No. aa1231036
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1231036 500 g
AA12310A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1231036 Supplier Thermo Scientific Chemicals Supplier No. 01231036
Only null left
Add to Cart

CAS: 10326-27-9 | BaCl2H4O2 | 244.26 g/mol

Barium chloride is usually employed as a chemical reagent to precipitate soluble sulfates and also in the purification of the seawater solution. It is practiced in several applications like heat-treating baths, waste water treatment, the production of PVC stabilizers, oil lubricants, fireworks materials, hardening of steel, in the manufacture of pigments and in the manufacture of other barium salts. It is applied to prepare composites to absorb ammonia and to increase the crystallinty of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Melting Point 113°C-2H2O
Physical Form Crystalline
Quantity 500 g
UN Number UN1564
Merck Index 14,971
Solubility Information Very soluble in water; Soluble in methanol; Insoluble in ethanol,acetone,ethyl acetate.
IUPAC Name barium(2+) dihydrate dichloride
Formula Weight 244.28 (208.25 Anhydrous)
Grade ACS
Density 3.097
Chemical Name or Material Barium chloride, dihydrate
Show More Show Less
missing translation for 'healthHazard1' H301-H319-H332
missing translation for 'healthHazard2' GHS H Statement
H301-H332
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 233-788-1
RTECSNumber CQ8751000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.