Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride Dihydrate, ACS Reagent, Reagents

Catalog No. 01338073
Change view
Click to view available options
Quantity:
100 mL
500 g
50 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-073 100 mL
01-338-070 500 g
01-338-069 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-338-073 Supplier Reagents Holdings Llc Supplier No. C1147000100A6
 Restricted Item
Add to Cart
Add to Cart

Barium Chloride Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym Barium dichloride
IUPAC Name barium(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Boiling Point 1560°C
Melting Point 963°C
Color Colorless/White
Physical Form Solid
Quantity 100 mL
Grade ACS
Packaging Poly Bottle
Specific Gravity 3.86
Chemical Name or Material Barium Chloride Dihydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.