Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride Solution (20% w/v), Fisher Chemical™

Catalog No. SB8224 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB822-4 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB822-4

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10361-37-2 , 7732-18-5
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+) dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Color Colorless
Physical Form Liquid
Quantity 4 L
Assay Percent Range 19.5 to 20.5 %
Percent Purity 19.5 to 20.5%
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Barium Chloride Solution, 20% w/v
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.