Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine, Reagents

Catalog No. 01335801
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-801 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-801 Supplier Reagents Holdings Llc Supplier No. CB1099001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1300-92-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless to Light Gray/Blue
Physical Form Liquid
Quantity 1 L
Percent Purity 0.1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.002
Chemical Name or Material Barium Diphenylamine Sulfonate, 0.1% w/v for Chlorine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.