Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium fluoride, 99.9% (metals basis)

Catalog No. AA3638014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3638014 25 g
AA3638022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3638014 Supplier Thermo Scientific Chemicals Supplier No. 03638014
Add to Cart

CAS: 7787-32-8 | BaF2 | 175.32 g/mol

Barium fluoride is used as a material to make optical components such as lenses, in windows for infrared spectroscopy, in particular in the field of fuel oil analysis, used as a preopacifying agent and in enamel and glazing frits production. It is also used in the production of welding agents, metallurgy, as a molten bath for refining aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-32-8
Molecular Formula BaF2
Molecular Weight (g/mol) 175.32
MDL Number MFCD00003450
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
PubChem CID 62670
SMILES [F-].[F-].[Ba++]

Specifications

Boiling Point 2260°C
Melting Point 1353°C
Color Colorless
Physical Form Fused Lump
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in methanol,ethanol,water 1.2g/L.
IUPAC Name barium(2+) difluoride
Formula Weight 175.36
Odor Odorless
Refractive Index 1.4741
Density 4.83
Chemical Name or Material Barium fluoride
Show More Show Less
missing translation for 'healthHazard1' H301-H332
missing translation for 'healthHazard3' P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 232-108-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.