Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium fluoride, ultra dry, 99.998% (metals basis)

Catalog No. AA3861018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3861018 50 g
AA3861009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3861018 Supplier Thermo Scientific Chemicals Supplier No. 03861018
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7787-32-8 | BaF2 | 175.32 g/mol

Barium Fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Barium Fluoride is used in spectroscopic components. Barium Fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-32-8
Molecular Formula BaF2
Molecular Weight (g/mol) 175.32
MDL Number MFCD00003450
InChI Key OYLGJCQECKOTOL-UHFFFAOYSA-L
Synonym barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
PubChem CID 62670
IUPAC Name barium(2+) difluoride
SMILES [F-].[F-].[Ba++]

Specifications

Boiling Point 2260°C
Melting Point 1353°C
Color White
Physical Form Powder
Quantity 50 g
UN Number UN1564
Merck Index 14,974
Solubility Information It is soluble in methanol,ethanol. Sparingly soluble in water.
Formula Weight 175.36
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.83 g/mL
Assay (metals basis)
Chemical Name or Material Barium fluoride, Ultra dry
Show More Show Less
Hazard Category H301-H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 232-108-0
RTECSNumber CQ9100000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.