Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide Crystals, Reagent Grade, Innovating Science™
SCI_ED

Catalog No. S000530 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000530 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger: Harmful if swallowed, causes severe skin burns and eye damage, harmful if inhaled.
Catalog No. S000530 Supplier Aldon Corporation Supplier No. IS11018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger: Harmful if swallowed, causes severe skin burns and eye damage, harmful if inhaled.

Chemical Identifiers

CAS 12230-71-6
Molecular Weight (g/mol) 315.48 g/mol

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 100 g
Packaging HDPE
Percent Purity 90 to 100 %
Grade Reagent
Chemical Name or Material Barium Hydroxide, Octahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.