accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OH2OH2OOHOHBa

CAS RN 12230-71-6

IUPAC Name: barium(2+) octahydrate dihydroxide
Synonyms: barium hydroxide octahydrate
Molecular Weight (g/mol): 315.46
Molecular Formula: BaH18O10
InChi Key: ZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
Molecular Weight (g/mol) 
  • (27)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
Grade 
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (2)
Percent Purity 
  • (8)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (1)
  • (6)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (6)
  • (13)
  • (5)
  • (1)
  • (27)
  • (2)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (7)
  • (2)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (6)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)