Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide Octahydrate, ACS, Ricca Chemical
SDP

Catalog No. RDCB0100500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCB0100500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCB0100500 Supplier Ricca Chemical Company Supplier No. RDCB0100500A6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
IUPAC Name barium(2+) octahydrate dihydroxide
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity min 98%
Grade ACS
Packaging Natural Poly Bottle
Appearance Colorless/White Solid
Chemical Name or Material Barium Hydroxide
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.