Learn More
Barium hydroxide octahydrate, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥98%, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Packaging | Plastic Bottle |
| Linear Formula | Ba(OH)2 · 8H2O |
| UN Number | 3262 |
| Formula Weight | 315.46g/mol |
| Percent Purity | ≥98% |
| Grade | puriss. p.a. |
| Chemical Name or Material | Barium Hydroxide Octahydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
| Hazard Statement | P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
| Precautionary Statement | H302 + H332-H314 |
| EINECSNumber | 241-234-5 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.