Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium(II) Bis(trifluoromethanesulfonyl)imide 99.0+%, TCI America™
SDP

Catalog No. B51711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51711G 1 g
B51715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51711G Supplier TCI America Supplier No. B51711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 168106-22-7
Molecular Formula C4H2BaF12N2O8S4
Molecular Weight (g/mol) 699.614
MDL Number MFCD22200529
InChI Key VJUKAFJLZPVWME-UHFFFAOYSA-N
Synonym barium bis trifluoromethanesulfonimide, barium bis-triflimide
PubChem CID 131725281
IUPAC Name barium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Ba]

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 697.60
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Barium(II) Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.